missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up, Maxi kit for gel extraction and PCR clean up
Description
- Two applications in one kit – one buffer with optimal performance for both applications
- High recoveries for fragments down to 50 bp
- Binding buffer with pH indicator
- Separate buffers for single stranded DNA or SDS containing samples available
- Suitable for all gel buffer systems (e.g., TAE, TBE)
- Ensures complete removal of contaminations such as: nucleoties, primers, enzymes, mineral oil, PCR additives, detergents, and dyes
- Typical downstream applications: cloning, sequencing, PCR, restriction analysis
Specifications
Specifications
| Format | Maxi Spin Column |
| Isolation Technology | Silica Membrane Technology |
| Sample Type | Agarose Gel, PCR Reaction Mixture |
| Efficiency | 70-95% recovery |
| For Use With (Equipment) | Centrifuge |
| For Use With (Application) | PCR Clean-up and Gel extraction |
| No. of Reactions | 20 |
| Purity | A260/A280: 1.75–1.85 |
| Sample Volume | <10 mL PCR reaction mixture, <10 g agarose gel |
| Size | 50 bp - 20 kbp (Fragment size) |
| Show More |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- P264-Wash with water thoroughly after handling.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?