missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Blood QuickPure Columns

Product Code. 15830928
Change view
Click to view available options
Quantity:
250 Pk.
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15830928 250 Pk. Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15830928 Supplier Macherey-Nagel™ Supplier No. 740569.250S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Replacement mini spin columns for use with NucleoSpin™ Blood QuickPure kits for ultra-fast purification of highly concentrated genomic DNA from blood and biological fluids.

NucleoSpin™ Blood QuickPure — For rapid purification of highly concentrated DNA from blood

  • Ultra-fast procedure
  • Washing and drying in one combined step (hands-on-time < 10 min)
  • Consistently high DNA yields
  • Elution volume only 30–50 μL
  • Highly concentrated DNA, ready to use for sensitive downstream applications

Specifications

Color Red ring
Format Mini Spin Column
Isolation Technology Silica Membrane Technology
Sample Type Body Fluids, Whole Blood (Human or Animal, Fresh or Frozen)
Elution Volume 30 to 50 μL
Final Product Type Highly concentrated DNA
For Use With (Equipment) NucleoSpin Blood QuickPure kit
For Use With (Application) Ultra-fast purification of genomic DNA from blood
Product Type Replacement Column
Quantity 250 Pk.
Sample Volume 5 to 200 μL
Yield 4 to 6 μg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.