missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ 96 Plant Core Kit, 96-well kit for DNA from plants
Description
contains Plant II binding plates, buffers, RNase A, 4 x 96 preps Macherey-Nagel
Specifications
Specifications
| Content And Storage | 18°C to 25°C |
| Format | 96-well Plate |
| Sample Type | Plants |
| For Use With (Application) | Isolation of DNA |
| No. of Reactions | 384 Preps |
| Sample Volume | 20–100 mg wet weight |
| Size | 50 bp–approx. 50 kbp |
| Target | DNA |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction