Learn More
Macherey-Nagel™ NucleoSpin™ 8 RNA Core Kit, 8-well kit for RNA purification
Product Code. 12728432
Change view
Click to view available options
Sample Size:
<2 x 106 cultured cells, <20 mg human/animal tissue
Unit Size:
384 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Size Unit Size
12728432 <2 x 106 cultured cells, <20 mg human/animal tissue 384 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12728432 Supplier Macherey-Nagel™ Supplier No. 740465.4
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
RNA isolation in flexible 8‐well strip format.
contains RNA binding strips, buffers, RNase-free rDNase, 48 x 8 preps Macherey-Nagel
Specifications
Sample Size <2 x 106 cultured cells, <20 mg human/animal tissue