Learn More
Macherey-Nagel™ Nucleosil™ EC Guard Columns
Product Code. 12738072
Click to view available options
Pore Size:
100Å
120Å
Unit Size:
Each
Pack of 3
Product Code. 12738072
Brand: Macherey-Nagel™ 721070.30
Specifications
Carbon Load 11%
pH 2 to 8
Particle Size 5 μm
Length (Metric) 4 mm
Pore Size 120Å
Type EC Guard Column
Diameter (Metric) Inner 3 mm
Stationary Phase NUCLEOSIL™ C18
Mobile Phase Compatibility Acetonitrile/Water
Solvents Acetonitrile/Water
Packing Material Silica
Crosslinking Octadecyl
Column Format Guard Column
For Use With (Application) RP chromatography
Technique HPLC
Particle Shape Spherical
Hydrophobicity Yes
Functionality/Interaction Type Hydrophobic
Endcapped Yes
USP Type L1