Learn More
Macherey-Nagel™ Nucleosil™ Chiral-2
Click to view available options
Diameter (Metric):
4 mm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) | 4 mm |
| For Use With | For Optical Resolution of D,L-Supidimide, Optical Resolution of D,L-Warfarin |
| Length (Metric) | 250 mm |
| Particle Size | 5 μm |
| Pore Size | 100 Å |
| USP Type | L36 |
| Type | Nucleosil Chiral-2 |