Learn More
Macherey-Nagel™ Nucleosil™ C18 Nautilus
Specifications
Specifications
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 250 mm |
| Particle Size | 5 μm |
| pH | 2 to 8 |
| Pore Size | 100 Å |
| USP Type | L60 |
| Carbon Load | 16% |
| Endcapped | Yes |
| Type | Nucleosil C18 Nautilus |