Learn More
Macherey-Nagel™ Nucleosil™ 120-5 C18 EC Column
Product Code. p-8012274
Click to view available options
Diameter (Metric):
4 mm
4.6 mm
Unit Size:
Each
Product Code. 10104651
Brand: Macherey-Nagel™ 720041.46
Only null left
Add to basket
Add to basket
Column Format: Analytical Column; Carbon Load: 11%; Pore Size: 120 Å; Column Phase: Reversed Phase
Nucleosil™ 120-5 C18 EC Column
Specifications
Column Chemistry Silica
Column Format Analytical Column
Crosslinking Octadecyl
Diameter (Metric) 4.6 mm
For Use With Analytical HPLC
Functionality/Interaction Type Hydrophobic
Hydrophobicity Yes
Length (Metric) 250 mm
Max. Pressure 5800 psi (400 bar)
Mobile Phase Compatibility Acetonitrile/water
Particle Size 5 μm
pH 2 to 8
Pore Size 120 Å
Quantity 1 Ea.
Solvents Acetonitrile/water
Surface Area 200 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Packing Material Silica
Carbon Load 11%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Type EC Column
Technique EC