Learn More
Macherey-Nagel™ Nucleosil™ C8 HD Reversed Phase HPLC Column, 5 μm, 4.6 mm x 150 mm
Product Code. 10155171
Click to view available options
Length (Metric):
150 mm
Unit Size:
Each
Product Code. 10155171
Brand: Macherey-Nagel™ 720194.46
Only null left
Add to basket
Add to basket
Column Format: Analytical Column; Carbon Load: 13%; Pore Size: 100 Å; Column Phase: Reversed Phase
Specifications
Column Chemistry Silica
Column Format Analytical Column
Crosslinking Octyl
Diameter (Metric) 4.6 mm
For Use With For HPLC, GC, TLC, SPE
Hydrophobicity Yes
Length (Metric) 150 mm
Max. Pressure 5800 psi (400 bar)
Mobile Phase Compatibility Acetonitrile/Water
Particle Shape Spherical
Particle Size 5 μm
pH 2 to 8
Pore Size 100 Å
Solvents 100-5 C8 HD
Surface Area 350 m²/g
Temperature 60°C
USP Type L7
Stationary Phase C8
Packing Material High Density Silica
Carbon Load 13%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Type EC Column
Technique Analytical HPLC