missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag Tissue Kit (DNA Purification from Cells and Tissue)

Product Code. 18844360
Change view
Click to view available options
Quantity:
384 Preps
96 Preps
Unit Size:
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18844360 96 Preps Pack of 96
18854360 384 Preps -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18844360 Supplier Macherey-Nagel™ Supplier No. 744330.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Streamlined magnetic-bead DNA purification from a wide range of cell and tissue samples using the NucleoMag Tissue Kit.

  • The NucleoMag Tissue kit combines the flexibility and versatility of the Tissue product series with magnetic bead-based processing.
  • Provides an automation-friendly workflow for DNA purification.
  • Uses a simple protocol that is effective for a broad range of sample types.
  • Compatible with a variety of open robotic platforms.

Specifications

Capacity 0.4 μg/μL Beads
Isolation Technology Magnetic Bead
Sample Type Cells, Human Tissue, Animal Tissue
Elution Volume 50 to 200 μL
Final Product Type DNA
For Use With (Application) Isolation of DNA
Product Line NucleoMag
Product Type NucleoMag Tissue Kit
Quantity 96 Preps
Purity 1.6 to 1.9
Sample Volume <20 mg Human/Animal Tissue, <106 Cells
Technique Magnetic Separation
Test Time 40 to 120 min./96 Preps
Yield 10 to 20 μg (20 mg tissue)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.