Learn More
Macherey-Nagel™ NucleoMag Dx Pathogen, CE-IVD certified kit for isolation of viral RNA
Product Code. 16682434
Change view
Click to view available options
Quantity:
384 Preps
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16682434 384 Preps 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16682434 Supplier Macherey-Nagel™ Supplier No. 744215.4
CE-IVD certified magnetic bead based kit for isolation of viral RNA from oral/nasal swabs and saliva (validated for SARS-CoV-2)
  • Intended use for human respiratory swabs and saliva
  • Magnetic bead based purification with proven automation concepts
Specifications
Concentration Depending on amount and quality of sample
Content And Storage 18°C to 25°C
Isolation Technology Magnetic Bead
Purification Time 40 to 120 min./96 preps
Elution Volume 50 μL to 100 μl
Sample Type Respiratory Swabs, Saliva (Human)
Final Product Type Viral RNA
For Use With (Application) NGS, RT-PCR
No. of Reactions 384 Preps
Quantity 384 Preps
Sample Size ≤200 μL swab wash solution/saliva
Size 300 bp (approx. 50 kbp)
Target Viral RNA
Volume (Metric) 0.4 μg/μL beads
Form Magnetic Bead
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoMag Dx Pathogen (4x96)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : guanidium thiocyanate, ethanol, gyanidium hydrochloride, tween 20, proteinase K