missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag DNA Forensic kit for DNA purification from forensic traces

Product Code. 15707317
Change view
Click to view available options
:
Quantity: 1 x 96 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15707317 Quantity: 1 x 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15707317 Supplier Macherey-Nagel™ Supplier No. 744660.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Flexible magnetic bead based isolation of DNA from forensic traces.

NucleoMag™ DNA Forensic

  • Excellent DNA purity from all casework samples
  • Uniform buffer system for high throughput analysis (NucleoMag™ DNA Forensic) and single sample analysis (NucleoSpin™ DNA Forensic)
  • Conformity to ISO 18385 for doubtless DNA profiling
  • Manual and automated isolation of DNA from casework samples, contact traces (e.g., dried blood spots, cigarette filters, swabs)
  • Applications: STR analysis, qPCR, any kind of enzymatic reaction

Specifications

Capacity 0.4 μg/μL beads
Concentration <1 ng/μL
Content And Storage Room temperature (18 - 25 °C)
Isolation Technology Magnetic Bead
Sample Type Casework Samples, Contact Traces
Efficiency Binding capacity: 0.4 μg/μL beads
Elution Volume Elution volume: 25 - 50 μL
For Use With (Equipment) Magnetic separator, separation plate
For Use With (Application) Isolation of genomic DNA from forensic samples
Includes NucleoMag F-Beads, buffers, liquid proteinase K, reducing agent TCEP
No. of Reactions 1 x 96 Preps
Yield 1 - 3 μg (from buccal swabs)
Form Highly Reactive Superparamagnetic Beads
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • NucleoMag DNA Forensic (1x96)
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H413-May cause long lasting harmful effects to aquatic life.
Supplemental information
  • MIXTURE LIST-Contains : guanidium hydrochloride, guanidium thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.