Learn More
Macherey-Nagel™ NucleoMag DNA Bacteria for DNA purification from bacteria and yeast
Product Code. 16475199
Change view
Click to view available options
No. of Reactions:
384 Preps
96 Preps
Unit Size:
384 reactions
96 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
16475199 96 Preps 96 reactions
16485199 384 Preps 384 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16475199 Supplier Macherey-Nagel™ Supplier No. 744310.1
  • ONLINE EXCLUSIVE PRICE
    279.00€
    276.15€ / 96 reactions
    Save 2.85€
    1% Off
 Restricted Item
In Stock
Add to basket
Add to basket
Magnetic bead based kit for the isolation of genomic DNA from bacteria and yeast.
  • Automation friendly solution for microbial samples
  • Environmentally sustainable buffer chemistry – no chaotropic salts
  • Combine with MN Bead Tubes for single sample processing or MN 96 Bead Plates for high throughput sample disruption
  • Liquid Proteinase K and Liquid RNase A for easy handling
Specifications
Capacity 0.4 μg/μL beads
Concentration Depending on amount and quality of sample
Content And Storage 18°C to 25°C
Isolation Technology Magnetic Bead
Sample Type Animal Tissue, Bacteria, Fungi, Human Tissue, Insects
Elution Volume 50 to 120 μL
For Use With (Application) Enzymatic Reactions, NGS, PCR, RT-PCR, Southern Blotting
No. of Reactions 96 Preps
Sample Volume <40 mg insects, <40 mg (108-109) bacteria, <30 mg fungi, <30 mg lipid rich tissue, <40 mg other tissues
Target DNA
Test Time 40 to 120 min/96 preps (excl. lysis)
Yield Depending on amount and quality of sample
Form Magnetic Beads