Learn More
Macherey-Nagel™ NucleoMag Plant kit for DNA purification from plants
Description
After lysis of the blood cells genomic DNA is selectively bound to the NucleoMag Blood beads. Contaminants are washed away and the DNA can be eluted from the beads under low-salt conditions.As the magnetic bead technology provides a closed system (no transfer of sample from one well to another is necessary during the preparation) the risk for cross-contamination is greatly reduced. The NucleoMag technology can be easily adapted to common laboratory automation platforms. It can be used either with static-pin separators (eg. NucleoMag SEP) or magnetic separators integrated into robotic workstations.Kit components: NucleoMag B-Beads, elution plate (u-bottom), buffers. For further information on this product please contact our Technical Support team.
- Magnetic bead technology
- Yield: 1μg to 4μg
- Sample material: 100μL whole blood
- Elution volume: 25μL to 100μL
- Binding capacity: 0.4μL/1μL beads
- Preparation time: 120 min./96 preps
- Fragment size: 300bp to 30kb
- Format: superparamagnetic beads (size of beads: 1μm to 3μm)
Specifications
Specifications
| Capacity | 0.4 μg/μL beads |
| Isolation Technology | Magnetic Bead |
| For Use With (Application) | For Manual and Automated Isolation of DNA |
| No. of Reactions | 24 x 96 |
| Product Type | NucleoMag™ 96 C-Bead |
| Test Time | 120min. |
| Yield | 10 to 20μg |
| Form | Highly Reactive Superparamagnetic Beads |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Respiratory sensitiser Category 1
- H225-Highly flammable liquid and vapour.
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contain: dodecyl sulfate, sodium salt / 2-propanol / ethanol, sodiumperchlorate