Learn More
Macherey-Nagel™ Nucleogel™ HPLC Column
Product Code. p-8012246
Click to view available options
Material:
Nucleogel SAX 1000-8
Unit Size:
Each
Product Code. 10286841
Brand: Macherey-Nagel™ 719469
Specifications
Base Material Polymer-based
Column Chemistry Polymer
For Use With (Application) Purification of Peptides, Large Proteins and Oligonucleotides, HPLC
Solvent Compatibility 0.1 M Na2SO4 + 0.2 % NaN3
Particle Size 8 μm
Stationary Phase Polymer-based
Type HPLC Column
Material Nucleogel SAX 1000-8
USP Type L23