Learn More
Macherey-Nagel™ Nucleogel™ HPLC Column
Product Code. 10206511
Click to view available options
Unit Size:
Each
Material:
Nucleogel SAX 1000-8
Product Code. 10206511
Brand: Macherey-Nagel™ 720094.40
Specifications
Column Capacity 50 μeq/g
For Use With (Application) HPLC, Separation of Inorganic Anions
Solvent Compatibility 2 mM potassium hydrogen phthalate buffer pH 5.6
Particle Size 10 μm
Type HPLC Column