Learn More
Macherey-Nagel™ Nucleodur™ HILIC HPLC CC Guard Columns
Product Code. 12625999
Click to view available options
Diameter (Metric) Inner:
3 mm
4 mm
Particle Size:
3 μm
5 μm
Unit Size:
Each
Pack of 3
Product Code. 12625999
Brand: Macherey-Nagel™ 761590.30
Only null left
Specifications
Carbon Load 7%
pH 2 to 8.5
Particle Size 5 μm
Length (Metric) 8 mm
Pore Size 110Å
Type CC Guard Column
Diameter (Metric) Inner 3 mm
Stationary Phase NUCLEODUR™ HILIC
Mobile Phase Compatibility Acetonitrile/Water
Solvents Acetonitrile/Water
Packing Material Silica
Crosslinking Ammonium/Sulfonic Acid
Column Format Guard Column
For Use With (Application) Analytical and preparative applications as well as LC/MS
Technique HPLC
Particle Shape Spherical