missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoBond™ Xtra Maxi Plus EF, Maxi kit for endotoxin-free plasmid DNA
Description
Features
- Endotoxin-free
- Plasmid DNA with less than 0.05 EU/μg for transfection of highly sensitive cells
- Patented endotoxin removal by additional washing step – no extra ice incubation necessary
- Optimized column design – Maxi prep in 50 min.
- Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
- Improved silica material – typical yields of 1,000μg (Maxi)
- NucleoBond™Xtra Plus EF kits contain NucleoBond™Finalizers to speed up DNA precipitation
- High-speed Version
- High-copy plasmids from E. coli
- Low-copy plasmids from E. coli (increased buffer volumes required)
Specifications
Specifications
| Isolation Technology | Silica Membrane Technology |
| No. of Reactions | 20 |
| Sample Volume | <600 mL (high copy), <1200 mL (low copy) |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contain: 2-propanol/ethanol (diluted < 20 %)/sodium hydroxide solution (diluted < 2 %), dodecyl sulfate, sodi
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?