missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra BAC kit for large construct plasmid DNA

Product Code. 12713401
Click to view available options
Quantity:
2 Columns
250 to 750 mL
Unit Size:
10 preps
Each
This item is not returnable. View return policy

Product Code. 12713401

Brand: Macherey-Nagel™ 740436.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2nd generation of anion exchanger for large construct plasmid DNA.

Specifications

No. of Reactions 10 Reactions
Product Type Xtra BAC
Quantity 250 to 750 mL
Sample Volume 250 to 750 mL
Size < 300kbp
Test Time 75 min.
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : ethanol (diluted < 20 %) / sodium hydroxide solution (diluted < 2 %), dodecyl sulfate, sodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.