Learn More
Macherey-Nagel™ NucleoBond™ PC 500, Maxi Kit for Transfection-grade Plasmid DNA
Click to view available options
No. of Reactions:
10 Preps
100 Preps
25 Preps
50 Preps
Unit Size:
10 reactions
100 reactions
25 reactions
50 reactions
Description
Features
- The classic anion exchanger for purification of transfection-grade plasmid DNA
- Fast and easy clarification of lysates with NucleoBond™ Folded Filters, no centrifugation required, no shearing forces
- NucleoBond™ Finalizers available as separate tool to avoid time consuming centrifugation for plasmid precipitation
- Proven anion-exchange technology – transfection-grade plasmid DNA
- Separate binding columns of NucleoBond™ PC kits available as NucleoBond™ AX columns
Applications:
- High-copy plasmids from E. coli
- Low-copy plasmids from E. coli (increased buffer volumes required)
Specifications
Specifications
| Format | Maxi |
| Isolation Technology | Anion Exchange Chromatography Technology |
| Final Product Type | Columns PC 500 |
| For Use With (Application) | High-copy/low-copy plasmids from E. coli |
| Includes | AX 500 Column, Buffers, RNase A, NucleoBond Folded Filters XL |
| No. of Reactions | 100 Preps |
| Product Type | Anion exchange column |
| Sample Volume | 30–150mL (HC) 100–500mL (LC) |
| Size | <300 kbp |
| Test Time | Prep time: 80–90 min./4 preps |
| Show More |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : ethanol (diluted < 20 %)/sodium hydroxide solution (diluted< 2 %)/dodecyl sulfate, sodium salt