Learn More
Macherey-Nagel™ NucleoBond™ Finalizer
Concentrate and desalinate plasmid and cosmid DNA eluates obtained by anion-exchange chromatography. The NucleoBond™ Finalizer is a quick, efficient alternative to centrifugation and perfect for all types of downstream applications.
57.00€ - 86.60€
Specifications
For Use With (Application) | Applications: Quick concentration and desalination of plasmid and cosmid DNA eluates obtained by anion-exchange chromatography, Typical downstream applications: transfection, in-vitro transcription, automated or manual sequencing, cloning, hybridization, and PCR. |
---|---|
Product Type | Desalting tool |
Size | 2-50kbp |
Test Time | 5min. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12710941
|
Macherey-Nagel™
740519.20 |
2 x 30, 2 x 1 mL |
57.00€
Pack of 20 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
11922442
|
Macherey-Nagel™
740520.20 |
20 x 30, 20 x 1 mL |
86.60€
Pack of 20 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
Description
Features
- Desalting tools for speeding up the time-consuming precipitation step after anion-exchange plasmid purification
- Eliminates centrifugation steps for precipitation – time saving from >1 h to only 5 minutes
- Two sizes available, to be combined with Midi and Maxi preparations
- Highly concentrated plasmid DNA
- No loss of DNA pellets or incomplete solubilization of hardly visible precipitates
Applications
- Quick concentration and desalination of plasmid and cosmid DNA eluates obtained by anion-exchange chromatography
- Typical downstream applications: transfection, in-vitro transcription, automated or manual sequencing, cloning, hybridization, and PCR
Specifications
Applications: Quick concentration and desalination of plasmid and cosmid DNA eluates obtained by anion-exchange chromatography, Typical downstream applications: transfection, in-vitro transcription, automated or manual sequencing, cloning, hybridization, and PCR. | |
Desalting tool | |
2-50kbp | |
5min. |