Learn More
Macherey-Nagel™ NucleoBond™ Finalizer
Concentrate and desalinate plasmid and cosmid DNA eluates obtained by anion-exchange chromatography.  The NucleoBond™ Finalizer is a quick, efficient alternative to centrifugation and perfect for all types of downstream applications.
Brand:  Macherey-Nagel™ 740520.20
Additional Details : Weight : 0.77000kg
 View more versions of this product
Product Code. 11922442
  • 86.60€ / Pack of 20
Estimated Shipment: 27-05-2024
Description
Features 
  • Desalting tools for speeding up the time-consuming precipitation step after anion-exchange plasmid purification
  • Eliminates centrifugation steps for precipitation – time saving from >1 h to only 5 minutes
  • Two sizes available, to be combined with Midi and Maxi preparations
  • Highly concentrated plasmid DNA
  • No loss of DNA pellets or incomplete solubilization of hardly visible precipitates
Applications
  • Quick concentration and desalination of plasmid and cosmid DNA eluates obtained by anion-exchange chromatography
  • Typical downstream applications: transfection, in-vitro transcription, automated or manual sequencing, cloning, hybridization, and PCR   
Specifications
Specifications
Applications: Quick concentration and desalination of plasmid and cosmid DNA eluates obtained by anion-exchange chromatography, Typical downstream applications: transfection, in-vitro transcription, automated or manual sequencing, cloning, hybridization, and PCR.
20 x 30, 20 x 1 mL
5min.
Desalting tool
2-50kbp