Learn More
Tocris Bioscience™ NU 7441
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
NU 7441 is a potent and selective DNA-PK inhibitor (IC50 = 14 nM). Selective for DNA-PK over a range of kinases including mTOR, PI 3-K, ATM and ATR. Potentiates the effects of doxorubicin (Cat. No. 2252) and etoposide (Cat. No. 1226) in vitro and etoposide in vivo. Also enhances CRISPR-Cas9-mediated homology-directed repair (HDR) efficiency 2 to 3-fold, and decreases nonhomologous end-joining (NHEJ) frequency ∽40%.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | NU 7441 |
| Purity | >99% |
| Molecular Weight (g/mol) | 413.49 |
| Inhibitors | NU 7441 |