missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NR2F6 (Human) Recombinant Protein
Human NR2F6 full-length ORF (BAG37465.1, 1 a.a. - 404 a.a.) recombinant protein with GST tag at N-terminal.
Brand: Abnova™ H00002063-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAMVTGGWGGPGGDTNGVDKAGGYPRAAEDDSASPPGAASDAEPGDEERPGLQVDCVVCGDKSSGKHYGVFTCEGCKSFFKRSIRRNLSYTCRSNRDCQIDQHHRNQCQYCRLKKCFRVGMRKEAVQRGRIPHSLPGAVAASSGSPPGSALAAVASGGDLFPGQPVSELIAQLLRAEPYPAAAGRFGAGGGAAGAVLGIDNVCELAARLLFSTVEWARHAPFFPELPVADQVALLRLSWSELFVLNAAQAALPLHTAPLLAAAGLHAAPMAAERAVAFMDQVRAFQEQVDKLGRLQVDSAEYGCLKAIALFTPDACGLSDPAHVESLQEKAQVALTEYVRAQYPSQPQRFGRLLLRLPALRAVPASLISQLFFMRLVGKTPIETLIRDMLLSGSTFNWPYGSGQ
Specifications
BAG37465.1 | |
nuclear receptor subfamily 2, group F, member 6 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NR2F6 | |
GST |
2063 | |
Wheat germ expression system | |
10 μg | |
EAR-2, EAR2, ERBAL2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |