Learn More
Abnova™ NPM1 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
1475.00€ - 2430.00€
Especificaciones
Content And Storage | Store at 4°C in the dark. |
---|---|
Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
Format | Tube |
For Use With (Application) | FISH-Ce |
Gene ID (Entrez) | 4869 |
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
16083475
|
Abnova™
FS0044.100UL |
100 μL |
1475.00€
100µL |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
16093475
|
Abnova™
FS0044.200UL |
200 μL |
2430.00€
200µL |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Especificaciones
Store at 4°C in the dark. | |
Tube | |
4869 | |
Liquid |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
FISH-Ce | |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |