Learn More
Melford NPG [4-Nitrophenyl-β-glucuronide
Product Code. 30018554
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018554 1 g 1g
30018553 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30018554 Supplier Melford Supplier No. N120301.0
 Restricted Item
Estimated Shipment: 28-10-2026
Spectrophotometric substrate for detection of B-D-Glucuronidase.
Spectrophotometric substrate for detection of B-D-Glucuronidase.
Specifications
Quantity 1 g