missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The TLR2 Cell Lysate is derived from mouse. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| Cell Line | Mouse TLR2 Transfected Cell Lysate |
| For Use With (Application) | Western Blot |
| Quantity | 180 μL |
| Storage Buffer | 1X Sample Buffer containing b-Mercaptoethanol, 1X SDS sample buffer: 60mM Tris-Hcl (pH 6.8), 10mM EDTA, 2 % SDS, 0.03% Bromophenol Blue, 10% glycerol, 0.3% b-mercaptoethanol |
| Host Species | Mouse |
| Content And Storage | Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?