Learn More
Novus Biologicals™ Raw 264.7 Whole Cell Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Description
The Raw 264.7 Whole Cell Lysate is derived from mouse. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Blood Lymph |
| Quantity | 0.1 mL |
| Storage Buffer | Standard RIPA buffer including protease and phosphatase inhibitors and 0.1% SDS diluted in 1X lithium dodecyl sulfate (LDS) sample buffer with bromophenol blue and 40-70% glycerol, pH 8.4. BME has not been added. |
| Cellular Fraction | Whole |
| Host Species | Mouse |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Denatured |