Learn More
Novus Biologicals™ pCL-Eco Retrovirus Packaging Vector
Applications: Retroviral Production
Brand: Novus Biologicals™ NBP2-29540
Description
The pCL-ECO packaging vector is a part of the RetroMax expression system and has been designed to maximize recombinant-retrovirus titers in a simple, efficient, and flexible experimental system. By introducing a retroviral vector into a sell expressing retroviral proteins, retroviral particles (virions) are shed into the culture medium at the rate of about 1 infectious particle/cell/day. Retrovirus tropism is determined at 3 levels. The first is simply a function of viral envelope protein, gp70. The envelope determines which cells the virus will enter. gp70 comes in three different flavors for gene therapy. Retroviruses obtained by cotransfection with pCL-Eco vector will infect mouse and rat cells, but not human cells.
- Species: Human, Mouse, Rat
- Specificity: Ectotropic (usually) (MoMuLV) Mouse and rat cells only (not human), Amphotropic (from 4070A MuLV) Most mammalian cells (but not hamster) 10A1 (MuLV) Most mammalian cells (including hamster)
- For large scale production, this plasmid may be amplified on LB plates containing 50µg/mL ampicillin
- Buffer: 10µg in 20µL 1x TE (10mM Tris, pH 7.5, 1mM EDTA)
Specifications
| 10µg in 20µL 1x TE (10mM Tris, pH 7.5, 1mM EDTA) | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| 10 μg | |
| pCL-Eco Retrovirus Packaging Vector |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.