missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PC-12 Whole Cell Lysate

Product Code. 18044504 Shop All Bio Techne Products
Click to view available options
Quantity:
0.25 mg
Unit Size:
0.25mg
This item is not returnable. View return policy

Product Code. 18044504

Brand: Novus Biologicals™ NBP213170

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The PC-12 Whole Cell Lysate is derived from rat. It has been validated for the following applications: Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Immunoprecipitation, Western Blot
Tissue Nervous
Quantity 0.25 mg
Storage Buffer Standard RIPA buffer including protease and phosphotase inhibitors and 0.1% SDS
Test Specificity PC-12 (rat adrenal gland pheochromocytoma) whole cell lysate (non-denatured)
Cellular Fraction Whole
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.