missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Nuclear Extraction Kit

Product Code. 18749403 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18749403 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18749403 Supplier Novus Biologicals™ Supplier No. NBP229447

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications: Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage is content dependent.
Quantity 1 Kit
Kit Contents 10X Hypotonic Lysis Buffer 10mL, 1X Nuclear Extraction Buffer 10mL, 10% Detergent Solution 10mL, 10X PBS 2 x 50mL, 1 M DTT (for Nuclear Extraction from tissue) 100μL, 100X Protease Inhibitor Cocktail (PIC) 100μL, 100 mM PMSF 10mL
For Use With (Application) Western Blotting
Product Type Nuclear Extraction Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.