missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Lano Antibody Blocking Peptide

Product Code. 18228295 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18228295 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18228295 Supplier R&D Systems™ Supplier No. NBP176678PEP
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide containing 15 amino acids near the carboxy terminus of human Lano. Source: Synthetic (Accession #: AAK72246) The Lano Blocking Peptide has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 55227
Species Human
Product Line Novus
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-76678. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias dJ523E19.1, FLJ10775, FLJ11834, LANOLANO adapter protein, LAP (leucine-rich repeats and PDZ) and no PDZ protein, LAP and no PDZ protein, leucine rich repeat containing 1, leucine-rich repeat-containing 1, leucine-rich repeat-containing protein 1
Gene Symbol LRRC1
Product Type Lano
Quantity 0.05 mg
Regulatory Status RUO
Source Synthetic
Termination 15 amino acids near the carboxy terminus of human Lano
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.