missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Jurkat Cell Lysate

Product Code. 18191315 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18191315 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18191315 Supplier Novus Biologicals™ Supplier No. NBP225046

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The Jurkat Cell Lysate is derived from human. It has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.1 mg
Storage Buffer 1X SDS sample buffer: 60mM Tris-HCl (pH 6.8) 10mM EDTA 2 % SDS 0.03% Bromophenol Blue 10% glycerol 0.3% Beta-mercaptoethanol
Host Species Human
Content And Storage -80°C in the dark. Avoid freeze-thaw cycles.

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.