missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human FXYD5/Dysadherin His Protein
Highly purified. Generating reliable and reproducible results.
Brand: Novus Biologicals™ NBP2-23064
This item is not returnable.
View return policy
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 22-145 of Human FXYD5/Dysadherin The Recombinant Human FXYD5/Dysadherin Protein is derived from E. coli. The Recombinant Human FXYD5/Dysadherin Protein has been validated for the following applications: SDS-Page.Specifications
| NP_001158077 | |
| 53827 | |
| FXYD5/Dysadherin Protein | |
| 0.1mg | |
| FXYD5, 22-145aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSQTLKDTT SSSSADSTIM DIQVPTRAPD AVYTELQPTS PTPTWPADET PQPQTQTQQL EGTDGPLVTD PETHKSTKAA HPTDDTTTLS ERPSPSTDVQ TDPQTLKPSG FHEDDPFFYD EHTLRKR | |
| RUO | |
| Yes |
| 20mM Tris-HCl buffer (pH 8.0), 0.1 M NaCl, 10% glycerol, 1mM DTT | |
| 16.1kDa | |
| SDS-PAGE | |
| E. Coli | |
| −20°C or −80°C. Avoid freeze-thaw cycles. | |
| Unlabeled |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?