Learn More
Novus Biologicals™ BSA Removal Antibody Purification Kit
Product Code. 18778803 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18778803 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18778803 Supplier Novus Biologicals™ Supplier No. 8200100
  • ONLINE EXCLUSIVE PRICE
    507.00€
    478.80€ / 1mL
    Save 28.20€
    5% Off
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Antibody purification kit
Common commercial BSA removal techniques can involve many laborious steps. The BSA Removal Kit is a simple 1 step, 10 minute method which effectively separates the BSA from the antibody and the BSA Removal Kit can be used on any antibody sub-type or species.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
Quantity 1 mL
Product Type BSA Removal Antibody Purification Kit