missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ BRSK1 Antibody Blocking Peptide

Product Code. 18219865 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18219865 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18219865 Supplier Novus Biologicals™ Supplier No. NBP176778PEP
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide containing 28 amino acids near the carboxy terminus of human BRSK1. Source: Synthetic (Accession #: Q8TDC3) The BRSK1 Blocking Peptide has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 84446
Species Human
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-76778. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias BR serine/threonine kinase 1, EC 2.7.11, EC 2.7.11.1, FLJ43009, KIAA1811BR serine/threonine-protein kinase 1, protein kinase SAD1A, SAD1, SAD1 kinase, Serine/threonine-protein kinase SAD-B
Gene Symbol BRSK1
Product Type BRSK1
Quantity 0.05 mg
Regulatory Status RUO
Source Synthetic
Termination 28 amino acids near the carboxy terminus of human BRSK1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.