Learn More
Monel™ 400 gauze, 80 mesh woven from 0.14mm (0.0055 in.) dia. wire, Thermo Scientific™
Click to view available options
Quantity:
150 x 150 mm
300 x 300 mm
Unit Size:
Each
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Chemical Name or Material | Monel™ 400 gauze |
| Name Note | 80 mesh woven from 0.14mm (0.0055 in.) dia. wire |
| Quantity | 300 x 300 mm |
| Mesh Size | 80 Mesh |