Learn More
Emma Safety Footwear Novara Safety Shoes
Product Code. 15998201
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15998201
Brand: Emma Safety Footwear MM13109039
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Classic Brogue design with little more ankle support thanks to its extended shank. Virtually impenetrable nitrile rubber soles combined with the shoe's sophisticated looks, allow the wearer to move easily between the office and warehouse.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather / Nubuck construction
  • Chemical resistant / Liquid repellant
  • Nitrile Rubber Heat Resistant Outsole
  • ESD certified
  • Chrome-free lining
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Brown
Design Novara
Liner Material Hydro-Tec™ Sanitized Silver & Wellness leather
Material Leather, Nubuck
Size 39
Toe Material Steel