missing translation for 'onlineSavingsMsg'
Learn More

Melford Nourseothricin Sulfate

Product Code. 30018588
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
This item is not returnable. View return policy

Product Code. 30018588

Brand: Melford N512001.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nourseothricin Sulfate is a broad-spectrum antibiotic.

Nourseothricin Sulfate is a broad-spectrum antibiotic. It is commonly used as a selection agent in molecular biology, genetics, and cell culture applications. Nourseothricin, also known as Streptothricin, is a natural product derived from Streptomyces species.

In molecular biology, Nourseothricin Sulfate is often employed as a selectable marker in gene expression studies and genetic engineering experiments. It works by inhibiting protein synthesis in bacterial and eukaryotic cells, leading to cell death.

Researchers use this antibiotic to introduce or maintain plasmids carrying resistance genes encoding Nourseothricin acetyltransferase (NAT) enzyme. The NAT enzyme inactivates Nourseothricin, allowing only the transformed cells to survive and propagate.

Nourseothricin Sulfate is also utilized in cell culture and microbiology. It can be used to selectively kill bacterial or fungal contaminants in cell cultures, thereby ensuring the purity of cell lines.

Additionally, it finds application in the isolation and maintenance of resistant strains in microbiological research.

Specifications

Chemical Name or Material Nourseothricin Sulfate
CAS 96736-11-7
Quantity 1 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.