Learn More
Cole-Parmer™ North Half-Mask Respirator
Product Code. 15226966 Shop All Cole-Parmer Products
Change view
Click to view available options
:
North Half-Mask Respirator; Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15226966 North Half-Mask Respirator; Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15226966 Supplier Cole-Parmer™ Supplier No. 8160612
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
North Half-Mask Respirator is made of soft, pliable, non-allergenic silicone offering excellent comfort and fit. Low-profile positioning on the face gives a wide field of vision and room for protective eyewear.
  • Wraparound lens provides over 200° field of vision
  • Superior comfort and fit
  • Contoured flange ensures proper seal
Specifications
Certifications/Compliance NIOSH
Color Black
Material Silicone
Product Type Half-Mask Respirator
Size Medium
Quantity 1 Ea.