missing translation for 'onlineSavingsMsg'
Learn More

Norneo Sildenafil, TRC

Product Code. 30011081
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011081 25 mg 25mg
30006665 5 mg 5mg
30008173 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011081 Supplier Toronto Research Chemicals Supplier No. TRCN82430025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371959-09-0
Molecular Formula C22 H29 N5 O4 S
Molecular Weight (g/mol) 459.56
Synonym Piperidine, 1-[[3-(4,7-dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]- (9CI), 5-[2-Ethoxy-5-(1-piperidinylsulfonyl)phenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one, CID 1896867, Norneosildenafil
IUPAC Name 5-(2-ethoxy-5-piperidin-1-ylsulfonylphenyl)-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
SMILES CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCCCC4

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H29N5O4S/c1-4-9-17-19-20(26(3)25-17)22(28)24-21(23-19)16-14-15(10-11-18(16)31-5-2)32(29,30)27-12-7-6-8-13-27/h10-11,14H,4-9,12-13H2,1-3H3,(H,23,24,28)
Formula Weight 459.194
Percent Purity >95%
Chemical Name or Material Norneosildenafil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.