Learn More
Norneo Sildenafil, TRC
Product Code. 30006665
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006665 5 mg 5mg
30011081 25 mg 25mg
30008173 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006665 Supplier Toronto Research Chemicals Supplier No. TRCN8243005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 371959-09-0
Molecular Formula C22 H29 N5 O4 S
Molecular Weight (g/mol) 459.56
Synonym Piperidine, 1-[[3-(4,7-dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]- (9CI), 5-[2-Ethoxy-5-(1-piperidinylsulfonyl)phenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one, CID 1896867, Norneosildenafil
IUPAC Name 5-(2-ethoxy-5-piperidin-1-ylsulfonylphenyl)-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
SMILES CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCCCC4
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C22H29N5O4S/c1-4-9-17-19-20(26(3)25-17)22(28)24-21(23-19)16-14-15(10-11-18(16)31-5-2)32(29,30)27-12-7-6-8-13-27/h10-11,14H,4-9,12-13H2,1-3H3,(H,23,24,28)
Formula Weight 459.194
Percent Purity >95%
Chemical Name or Material Norneosildenafil
Grade HPLC