Learn More
Norglipin, TRC
Specifications
Specifications
| Chemical Name or Material | (1R,3r,5S)-8-azabicyclo(3.2.1)oct-3-yl hydroxydiphenylacetate |
| CAS | 16444-19-2 |
| Molecular Formula | C21 H23 N O3 |
| Quantity | 10 mg |
| Synonym | Trospium Imp. B, ÃŽ±-hydroxy-ÃŽ±-phenyl-benzeneacetic acid (3-endo)-8-azabicyclo[3.2.1]oct-3-yl ester, (1R,3r,5S)-8-azabicyclo[3.2.1]oct-3-yl hydroxydiphenylacetate, 1ÃŽ±H,5ÃŽ±H-Nortropan-3ÃŽ±-ol','benzilate (ester) (8CI), Benzeneacetic acid','ÃŽ±-hydroxy-ÃŽ±-phenyl-','8-azabicyclo[3.2.1]oct-3-yl ester','endo-, Benzilic acid','1ÃŽ±H,5ÃŽ±H-nortropan-3ÃŽ±-yl ester (8CI), Norglipin |
| InChI Formula | InChI=1S/C21H23NO3/c23-20(25-19-13-17-11-12-18(14-19)22-17)21(24,15-7-3-1-4-8-15)16-9-5-2-6-10-16/h1-10,17-19,22,24H,11-14H2/t17-,18+,19+ |
| SMILES | OC(C(=O)O[C@H]1C[C@H]2CC[C@@H](C1)N2)(c3ccccc3)c4ccccc4 |
| IUPAC Name | [(1S,5R)-8-azabicyclo[3.2.1]octan-3-yl] 2-hydroxy-2,2-diphenylacetate |
| Molecular Weight (g/mol) | 337.41 |
| Formula Weight | 337.1678 |