Learn More
Norethynodrel (>90%), TRC
Product Code. 18060440
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18060440 25 mg 25mg
18012150 2.5 mg 2.5mg
18033440 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18060440 Supplier Toronto Research Chemicals Supplier No. TRCN67602025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 68-23-5
Molecular Formula C20 H26 O2
Molecular Weight (g/mol) 298.42
Synonym 19-Nor-17α-pregn-5(10)-en-20-yn-3-one, 17-hydroxy- (6CI,8CI), 13-Methyl-17-ethynyl-17-hydroxy-1,2,3,4,6,7,8,9,11,12,13,14,16,17-tetradecahydro-15H-cyclopenta[a]phenanthren-3-one, 17-Hydroxy-19-nor-17α-pregn-5(10)-en-20-yn-3-one, 17α-Ethinyl-17β-hydroxy-δ5(10)-estren-3-one, 17α-Ethynyl-17-hydroxy-5(10)-estren-3-one, 17α-Ethynyl-17β-hydroxy-5(10)-estrene-3-one, 17α-Ethynyl-17β-hydroxy-δ5(10)-estren-3-one, 17α-Ethynylestr-5(10)-ene-17β-ol-3-one, 17α-Ethynylestra-5(10)-ene-17β-ol-3-one, 17β-Hydroxy-19-nor-17α-pregn-5(10)-en-20-yn-3-one, 3-Oxo-17β-hydroxy-17α-ethynyl-5(10)-estrene, Enidrel, NSC 15432, Norethinodrel, Norethynodrel, Noretynodrel, SC 4642, 19-Norpregn-5(10)-en-20-yn-3-one, 17-hydroxy-, (17α)-
IUPAC Name (8R,9S,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,4,6,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=C3CCC(=O)C4)[C@@H]1CC[C@@]2(O)C#C
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C20H26O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,16-18,22H,4-12H2,2H3/t16-,17-,18+,19+,20+/m1/s1
Formula Weight 298.1933
Chemical Name or Material Norethynodrel