Learn More
(±)-Norepinephrine-2,5,6,alpha,beta,beta-d6 HCl, CDN
Product Code. 30317459
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30317459
Brand: CDN CDND66340.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (±)-Norepinephrine-D6 Hydrochloride (Noradrenaline)
CAS 1219803-04-9
Molecular Formula C8 2H6 H5 N O3 . Cl H
Quantity 0.005 g
Synonym (±)-Norepinephrine-D6 hydrochloride, DL-Norepinephrine-d6 Hydrochloride
InChI Formula InChI=1 S/C8H11NO3.ClH/c9-4-8(12)5-1-2-6(10)7(11)3-5;/h1-3,8,10-12 H,4,9H2;1 H/i1D,2 D,3 D,4D2,8 D;
SMILES Cl.[2 H]c1c([2 H])c(c([2 H])c(O)c1O)C([2 H])(O)C([2 H])([2 H])N
IUPAC Name 4-(2-amino-1,2,2-trideuterio-1-hydroxyethyl)-3,5,6-trideuteriobenzene-1,2-diol;hydrochloride
Molecular Weight (g/mol) 211.68
Formula Weight 211.0882 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity