Learn More
Nordihydrocapsaicin, TRC
Product Code. 30004581
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004581 250 mg 250mg
18030750 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30004581 Supplier Toronto Research Chemicals Supplier No. TRCN672600250MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28789-35-7
Molecular Formula C17H27NO3
Molecular Weight (g/mol) 293.4
Synonym Octanamide, N-[(4-hydroxy-3-methoxyphenyl)methyl]-7-methyl-, Octanamide, 7-methyl-N-vanillyl- (8CI), N-[(4-Hydroxy-3-methoxyphenyl)methyl]-7-methyloctanamide, Nordihydrocapsaicin, Norhydrocapsaicin
IUPAC Name N-[(4-hydroxy-3-methoxyphenyl)methyl]-7-methyloctanamide
SMILES COc1cc(CNC(=O)CCCCCC(C)C)ccc1O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C17H27NO3/c1-13(2)7-5-4-6-8-17(20)18-12-14-9-10-15(19)16(11-14)21-3/h9-11,13,19H,4-8,12H2,1-3H3,(H,18,20)
Formula Weight 293.1991
Percent Purity >95%
Chemical Name or Material Nordihydrocapsaicin
Grade HPLC