Learn More
Emma Safety Footwear Nordic Oiled Nubuck Leather D-Fit Shoes
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Each
Description
- Oil Nubuck uppers
- Hydro-Tec™ sanitized silver lining
- Non-woven antistatic insole
- GripForce™ Easy Twist outsole
Specifications
Specifications
| Color | Black, Brown |
| Liner Material | Hydro-Tec sanitized silver |
| Material | Leather |
| Size | 42 |
| Sole Features | Non-woven antistatic insole |
| Sole Material | GripForce™ Easy Twist outsole |
| Toe Material | Steel |
| Product Type | Oiled Nubuck Leather Shoes |