Learn More
Thermo Scientific™ Nonsterile Pipette Tips for Beckman™ Liquid Handling Systems
Shop All MBP Molecular BioProducts Products
Description
Increase pipetting performance through the superior tip straightness and low %CV of these pipette tips for automated liquid handling systems.
Increase pipetting performance with Thermo ScientificTM Nonsterile Pipette Tips for BeckmanTM Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. They provide accurate and precise performance because of superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
- Available in low retention and extended length
- Lot-tested during production on actual workstation for which they are created
- Extended length tips compatible with 96-, 384- and 1536- DeepWell™ plates
- Low percentage coefficient of variation (%CV)
Specifications
Specifications
| Volume (Metric) | 20 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Format | Rack |
| Tip Style | Conductive |
| For Use With (Equipment) | Beckman Coulter Biomek FX, Beckman Coulter Biomek NX SPAN-8 |
| Product Line | Beckman |
| Fit for Purpose | Certified Quality |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.