missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonanoic-d17 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | n-Nonanoic Acid-d17 |
| CAS | 130348-94-6 |
| Molecular Formula | C9 D17 H O2 |
| Quantity | 0.5 g |
| Synonym | 1-Octanecarboxylic Acid-d17, Emery 1203-d17, Grantrico-d17, NSC 62787-d17, NSC 65450-d17, Nonanoic-d17 acid (9 CI), Nonanoic-d17 acid (9 CI), Nonanoic-d17 acid (9 CI), Nonanoic-d17 acid (9 CI), Nonanoic-d17 acid (9 CI), Nonanoic-d17 acid (9 CI), Nonanoic-d17 acid (9 CI), Nonanoic-d17 acid (9 CI), Nonanoic-d17 acid (9 CI), Nonanoic-d17 acid (9 CI), Nonanoic-d17 acid (9 CI) |
| InChI Formula | InChI=1 S/C9H18O2/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3,(H,10,11)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecadeuteriononanoic acid |
| Molecular Weight (g/mol) | 175.34 |
| Formula Weight | 175.2374 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction