Learn More
Nonanoic-9,9,9-d3 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | n-Nonanoic-9,9,9-d3 acid |
| CAS | 134646-27-8 |
| Molecular Formula | C9 D3 H15 O2 |
| Quantity | 0.25 g |
| Synonym | Nonanoic-9,9,9-d3 acid (9 CI), Nonanoic-9,9,9-D3 acid, 1-Octanecarboxylic-8,8,8-d3 acid, n-Nonanoic acid-d3, Pelargonic acid-ω-d3, Nonanoic acid-d3 |
| InChI Formula | InChI=1 S/C9H18O2/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3,(H,10,11)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])CCCCCCCC(=O)O |
| IUPAC Name | 9,9,9-trideuteriononanoic acid |
| Molecular Weight (g/mol) | 161.256 |
| Formula Weight | 161.15 g/mol |
| Show More |